Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(2,3,6,7-tetrahydro-2,6-dioxo-1-propyl-1H-purin-8-yl)-benzenesulfonic acid | 152529-79-8 | ≥98.0% | 350.35 g/mol | C14H14N4O5S | 1 ML
SDP

Catalog No. 5000247280
Encompass_Preferred

PSB 1115 is a selective A2B adenosine receptor antagonist used as a research tool to block A2B-mediated signaling in pharmacology studies. It is supplied as a concentrated solution for laboratory assays and analytical applications.

  • Selective A2B adenosine receptor antagonist
  • Supplied as 10 mM solution, 1 mL
  • Purity ≥98.0% (HPLC)
  • Molecular weight 350.35 g/mol
  • Molecular formula C14H14N4O5S
  • Water-soluble and suitable for in vitro and in vivo studies

Catalog No. 50-002-47280 Supplier Medchemexpress LLC Supplier No. HY10318210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSB 1115 is a selective A2B adenosine receptor antagonist used as a research tool to block A2B-mediated signaling in pharmacology studies. It is supplied as a concentrated solution for laboratory assays and analytical applications.

  • Selective A2B adenosine receptor antagonist
  • Supplied as 10 mM solution, 1 mL
  • Purity ≥98.0% (HPLC)
  • Molecular weight 350.35 g/mol
  • Molecular formula C14H14N4O5S
  • Water-soluble and suitable for in vitro and in vivo studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.