Learn More
Medchemexpress LLC 4-[[(2,3-dihydro-1,1,3,3-tetramethyl-2-oxo-1H-inden-5-yl)carbonyl]amino]-benzoic acid | 215307-86-1 | MFCD18384955 | 98.9% | 351.40 Da | C21H21NO4 | 5 MG
SDP
Catalog No. 5000239783
Encompass_Preferred
BMS 753 (CAS 215307-86-1) is an isotype-selective retinoic acid receptor α (RARα) agonist with a reported Ki of 2 nM. Supplied as a research-grade powder, it has molecular formula C21H21NO4 and molecular weight 351.40 Da, and is provided with high purity for laboratory use. The compound is intended for in vitro pharmacology and receptor signaling studies and requires low-temperature storage when in solvent.
Catalog No. 50-002-39783 Supplier Medchemexpress LLC Supplier No. HY1073955MG
BMS 753 (CAS 215307-86-1) is an isotype-selective retinoic acid receptor α (RARα) agonist with a reported Ki of 2 nM. Supplied as a research-grade powder, it has molecular formula C21H21NO4 and molecular weight 351.40 Da, and is provided with high purity for laboratory use. The compound is intended for in vitro pharmacology and receptor signaling studies and requires low-temperature storage when in solvent.