Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[[(2,3-dihydro-1,1,3,3-tetramethyl-2-oxo-1H-inden-5-yl)carbonyl]amino]-benzoic acid | 215307-86-1 | MFCD18384955 | 98.9% | 351.40 g·mol⁻¹ | C21H21NO4 | 100 MG
SDP

Catalog No. 5000240882
Encompass_Preferred

BMS 753 is a research-grade, isotype-selective retinoic acid receptor α (RARα) agonist reported to have a Ki of 2 nM. It is supplied as a solid research chemical with documented solubility and storage recommendations for in vitro and in vivo use.

  • Potent RARα agonist with reported Ki = 2 nM.
  • High purity, reported ~98.9% by HPLC.
  • Molecular weight 351.40 g·mol⁻¹; formula C21H21NO4.
  • Soluble in DMSO for in vitro use (50 mg/mL) with recommended vehicle protocols for in vivo dosing.
  • Powder stability: -20°C recommended for long-term storage.
  • Supplied in multiple pack sizes suitable for assay development and preclinical studies.

Catalog No. 50-002-40882 Supplier Medchemexpress LLC Supplier No. HY107395100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS 753 is a research-grade, isotype-selective retinoic acid receptor α (RARα) agonist reported to have a Ki of 2 nM. It is supplied as a solid research chemical with documented solubility and storage recommendations for in vitro and in vivo use.

  • Potent RARα agonist with reported Ki = 2 nM.
  • High purity, reported ~98.9% by HPLC.
  • Molecular weight 351.40 g·mol⁻¹; formula C21H21NO4.
  • Soluble in DMSO for in vitro use (50 mg/mL) with recommended vehicle protocols for in vivo dosing.
  • Powder stability: -20°C recommended for long-term storage.
  • Supplied in multiple pack sizes suitable for assay development and preclinical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.