Learn More
Medchemexpress LLC 4-(2,3-dihydro-1,4-benzodioxin-6-yl)-3-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-1H-1,2,4-triazol-5-one | 883065-90-5 | >98.0% | 379.37 g/mol | C13H9N5O5S2 | 50 MG
SDP
Catalog No. 5000245040
Encompass_Preferred
BI-78D3 is a small-molecule, substrate-competitive inhibitor of c-Jun N-terminal kinase (JNK) used as a research tool to probe JNK signaling. It inhibits JNK kinase activity with a reported IC50 of 280 nM. The compound has molecular formula C13H9N5O5S2, molecular weight 379.37 g/mol, and CAS 883065-90-5. Recommended storage: powder at -20°C (long term) or 4°C (short term); store solutions at -80°C or -20°C depending on duration.
  • Substrate-competitive JNK inhibitor (IC50 280 nM).
  • Suitable for biochemical and cell-based studies of JNK signaling.
  • Molecular weight 379.37 g/mol; chemical formula C13H9N5O5S2.
  • Powder form stable for long-term storage at -20°C.
  • Available in small research pack sizes for assay development.
Catalog No. 50-002-45040 Supplier Medchemexpress LLC Supplier No. HY1036650MG
BI-78D3 is a small-molecule, substrate-competitive inhibitor of c-Jun N-terminal kinase (JNK) used as a research tool to probe JNK signaling. It inhibits JNK kinase activity with a reported IC50 of 280 nM. The compound has molecular formula C13H9N5O5S2, molecular weight 379.37 g/mol, and CAS 883065-90-5. Recommended storage: powder at -20°C (long term) or 4°C (short term); store solutions at -80°C or -20°C depending on duration.
  • Substrate-competitive JNK inhibitor (IC50 280 nM).
  • Suitable for biochemical and cell-based studies of JNK signaling.
  • Molecular weight 379.37 g/mol; chemical formula C13H9N5O5S2.
  • Powder form stable for long-term storage at -20°C.
  • Available in small research pack sizes for assay development.