Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-[2-(3-Pyridyl)-4-thiazolyl]biphenyl-4-boronic acid, 97%, Thermo Scientific™

Catalog No. AAH5835206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5835206 5 g
AAH5835203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5835206 Supplier Thermo Scientific Chemicals Supplier No. H5835206

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1449132-90-4
Molecular Formula C20H15BN2O2S
Molecular Weight (g/mol) 358.222
MDL Number MFCD22682966
InChI Key DPNCWVXNIXFYHI-UHFFFAOYSA-N
Synonym 4'-2-pyridin-3-yl-1,3-thiazol-4-yl-1,1'-biphenyl-4-ylboronic acid, 4'-2-3-pyridyl-4-thiazolyl biphenyl-4-boronic acid
PubChem CID 99738172
IUPAC Name [4-[4-(2-pyridin-3-yl-1,3-thiazol-4-yl)phenyl]phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CSC(=N3)C4=CN=CC=C4)(O)O

Specifications

Quantity 5 g
Formula Weight 359.23
Percent Purity 97%
Chemical Name or Material 4'-[2-(3-Pyridyl)-4-thiazolyl]biphenyl-4-boronic acid
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.