Learn More
Medchemexpress LLC 4-[2-[4-[3-(4-chlorophenyl)-2-propenyl]-1-piperazinyl]ethyl]benzoic acid sulfate | 86621-94-5 | MFCD07370131 | 98.2% | C22H27ClN2O6S | 10MG
SDP
Catalog No. 5000259106
Encompass_Preferred
BM 15766 sulfate is a small-molecule hypolipidemic agent that inhibits 7-dehydrocholesterol δ7-reductase and has been used in research to reduce plasma cholesterol levels and study cholesterol biosynthesis.
  • Selective inhibitor of 7-dehydrocholesterol δ7-reductase.
  • Demonstrated to reduce plasma cholesterol levels in model systems.
  • High purity suitable for biochemical assays (≈98.2%).
  • Provided as a sulfate salt to aid solubility and handling.
  • Available in small pack sizes for laboratory research use.
Catalog No. 50-002-59106 Supplier Medchemexpress LLC Supplier No. HY13731810MG
BM 15766 sulfate is a small-molecule hypolipidemic agent that inhibits 7-dehydrocholesterol δ7-reductase and has been used in research to reduce plasma cholesterol levels and study cholesterol biosynthesis.
  • Selective inhibitor of 7-dehydrocholesterol δ7-reductase.
  • Demonstrated to reduce plasma cholesterol levels in model systems.
  • High purity suitable for biochemical assays (≈98.2%).
  • Provided as a sulfate salt to aid solubility and handling.
  • Available in small pack sizes for laboratory research use.