Learn More
Medchemexpress LLC 4-(2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethyl-7-((3S)-3-piperidinylmethyl)oxy)-2-methyl-3-butyn-2-ol | 937174-76-0 | MFCD14105605 | 98.5% | 425.48 g/mol | C21H27N7O3 | 25 MG
SDP
Catalog No. 5000245834
Encompass_Preferred
GSK-690693 is a small-molecule, ATP-competitive pan-Akt inhibitor used for biochemical and cell-based research into PI3K/Akt signaling. It inhibits Akt1, Akt2, and Akt3 with low nanomolar potency (IC50s: 2 nM, 13 nM, 9 nM). The compound has molecular formula C21H27N7O3 and a molecular weight of 425.48 g/mol, and is supplied as a high-purity solid for laboratory research.
  • Inhibits Akt1, Akt2, and Akt3 with low nanomolar potency.
  • ATP-competitive mechanism of action.
  • Suitable for biochemical and cell-based assays.
  • Supplied as a high-purity solid (~98.5%).
  • Molecular formula C21H27N7O3; molecular weight 425.48 g/mol.
Catalog No. 50-002-45834 Supplier Medchemexpress LLC Supplier No. HY1024925MG
GSK-690693 is a small-molecule, ATP-competitive pan-Akt inhibitor used for biochemical and cell-based research into PI3K/Akt signaling. It inhibits Akt1, Akt2, and Akt3 with low nanomolar potency (IC50s: 2 nM, 13 nM, 9 nM). The compound has molecular formula C21H27N7O3 and a molecular weight of 425.48 g/mol, and is supplied as a high-purity solid for laboratory research.
  • Inhibits Akt1, Akt2, and Akt3 with low nanomolar potency.
  • ATP-competitive mechanism of action.
  • Suitable for biochemical and cell-based assays.
  • Supplied as a high-purity solid (~98.5%).
  • Molecular formula C21H27N7O3; molecular weight 425.48 g/mol.