Learn More
Medchemexpress LLC 4-(2-(4-(benzyloxy)phenyl)acetamido)-1-(3'-chloro-[1,1'-biphenyl]-4-carbonyl)piperidine-4-COOH | 100.0% | 583.07 g/mol | C34H31ClN2O5 | 10 MG
SDP
Catalog No. 5000287705
Encompass_Preferred
YL-365 is a small-molecule GPR34 agonist used in preclinical research on pain and cancer. It has molecular formula C34H31ClN2O5, a molecular weight of 583.07 g/mol, and an analytical purity of 99.96%. The compound is supplied as a solid in milligram quantities and should be stored at 4°C protected from light; in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month.
  • Acts as a GPR34 agonist to probe lysophosphatidylserine receptor signaling.
  • Suitable for pain and cancer preclinical studies.
  • High analytical purity for consistent experimental outcomes.
  • Available in small milligram quantities for dosing flexibility.
  • Stable when stored under recommended conditions and protected from light.
Catalog No. 50-002-87705 Supplier Medchemexpress LLC Supplier No. HY15681510MG
YL-365 is a small-molecule GPR34 agonist used in preclinical research on pain and cancer. It has molecular formula C34H31ClN2O5, a molecular weight of 583.07 g/mol, and an analytical purity of 99.96%. The compound is supplied as a solid in milligram quantities and should be stored at 4°C protected from light; in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month.
  • Acts as a GPR34 agonist to probe lysophosphatidylserine receptor signaling.
  • Suitable for pain and cancer preclinical studies.
  • High analytical purity for consistent experimental outcomes.
  • Available in small milligram quantities for dosing flexibility.
  • Stable when stored under recommended conditions and protected from light.