Learn More
Medchemexpress LLC 4-[[2-[(4-bromophenyl)thio]acetyl]amino]-1-(2-phenylethyl)-1H-pyrazole-3-carboxylic acid | 1021497-97-1 | MFCD30738005 | >=98.0% | 460.34 g/mol | C20H18BrN3O3S | 25 MG
SDP
Catalog No. 5000211678
Encompass_Preferred
KR-33493 is a small-molecule inhibitor of Fas-associated factor 1 (FAF1) that blocks Fas-mediated cell death and is used as a research tool to probe TNF receptor-related apoptosis pathways and evaluate neuroprotective mechanisms in preclinical studies.
  • Potent inhibition of Fas-mediated cell death.
  • Useful probe for apoptosis and TNF receptor pathway studies.
  • Applicable in preclinical neuroprotection research.
  • Stable as a powder when stored at low temperatures.
  • Provided in small milligram quantities suitable for laboratory assays.
Catalog No. 50-002-11678 Supplier Medchemexpress LLC Supplier No. HY10075525MG
Add to Cart
Add to Cart
KR-33493 is a small-molecule inhibitor of Fas-associated factor 1 (FAF1) that blocks Fas-mediated cell death and is used as a research tool to probe TNF receptor-related apoptosis pathways and evaluate neuroprotective mechanisms in preclinical studies.
  • Potent inhibition of Fas-mediated cell death.
  • Useful probe for apoptosis and TNF receptor pathway studies.
  • Applicable in preclinical neuroprotection research.
  • Stable as a powder when stored at low temperatures.
  • Provided in small milligram quantities suitable for laboratory assays.