Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[2-(4-nitro-1H-imidazol-1-yl)ethyl]morpholine | 6497-78-5 | MFCD02029312 | >98.0% | 226.24 | C9H14N4O3 | 5 MG
SDP

Catalog No. 5000242804
Encompass_Preferred

RGW-611 (4-[2-(4-nitro-1H-imidazol-1-yl)ethyl]morpholine) is a morpholine derivative used in laboratory research; it has been reported to enhance radiation-induced cell death in hypoxic cell models and to stimulate fatty acid synthesis. The compound is supplied in small quantities for in vitro studies and requires controlled storage to maintain stability.

  • Chemical name: 4-[2-(4-nitro-1H-imidazol-1-yl)ethyl]morpholine.
  • CAS number: 6497-78-5.
  • Molecular formula: C9H14N4O3.
  • Molecular weight: 226.24 g/mol.
  • Purity: >98.0% (HPLC).
  • Available sizes: 1 mg, 5 mg, 10 mg.
  • Storage: 4°C, protect from light; in solvent store at -80°C for long-term.

Catalog No. 50-002-42804 Supplier Medchemexpress LLC Supplier No. HY1078405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RGW-611 (4-[2-(4-nitro-1H-imidazol-1-yl)ethyl]morpholine) is a morpholine derivative used in laboratory research; it has been reported to enhance radiation-induced cell death in hypoxic cell models and to stimulate fatty acid synthesis. The compound is supplied in small quantities for in vitro studies and requires controlled storage to maintain stability.

  • Chemical name: 4-[2-(4-nitro-1H-imidazol-1-yl)ethyl]morpholine.
  • CAS number: 6497-78-5.
  • Molecular formula: C9H14N4O3.
  • Molecular weight: 226.24 g/mol.
  • Purity: >98.0% (HPLC).
  • Available sizes: 1 mg, 5 mg, 10 mg.
  • Storage: 4°C, protect from light; in solvent store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.