Learn More
Medchemexpress LLC 4-(2-(4-nitro-1H-imidazol-1-yl)ethyl)morpholine | 6497-78-5 | MFCD02029312 | >98.0% | 226.24 g/mol | C9H14N4O3 | 25 MG
SDP
Catalog No. 5000244493
Encompass_Preferred
RGW-611 (4-(2-(4-nitro-1H-imidazol-1-yl)ethyl)morpholine) is a morpholine-derived research compound reported to sensitize hypoxic cells to radiation and to stimulate fatty acid synthesis. Supplied as a solid for laboratory research use.
  • Enhances radiation-induced cell death in hypoxic V79-379A cells.
  • Stimulates fatty acid synthesis in cellular assays.
  • High purity suitable for research (>98.0% by HPLC).
  • Molecular formula C9H14N4O3; molecular weight 226.24 g/mol.
  • Store protected from light; use cold storage for stock solutions.
Catalog No. 50-002-44493 Supplier Medchemexpress LLC Supplier No. HY10784025MG
RGW-611 (4-(2-(4-nitro-1H-imidazol-1-yl)ethyl)morpholine) is a morpholine-derived research compound reported to sensitize hypoxic cells to radiation and to stimulate fatty acid synthesis. Supplied as a solid for laboratory research use.
  • Enhances radiation-induced cell death in hypoxic V79-379A cells.
  • Stimulates fatty acid synthesis in cellular assays.
  • High purity suitable for research (>98.0% by HPLC).
  • Molecular formula C9H14N4O3; molecular weight 226.24 g/mol.
  • Store protected from light; use cold storage for stock solutions.