Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2,6-Dichloro-4-pyrimidyl)morpholine 98.0+%, TCI America™
SDP

Catalog No. D45715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45715G 5 g
D45711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45715G Supplier TCI America Supplier No. D45715G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 52127-83-0 | Formula: C8H9Cl2N3O | Weight: 234.08 g/mol

Chemical Identifiers

CAS 52127-83-0
Molecular Formula C8H9Cl2N3O
Molecular Weight (g/mol) 234.08
MDL Number MFCD09746259
InChI Key QGGYMWHOBGSQCF-UHFFFAOYSA-N
Synonym 2,4-Dichloro-6-(4-morpholino)pyrimidine
PubChem CID 11586699
IUPAC Name 4-(2,6-dichloropyrimidin-4-yl)morpholine
SMILES C1COCCN1C2=CC(=NC(=N2)Cl)Cl

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 5 g
Formula Weight 234.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2,6-Dichloro-4-pyrimidyl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.