Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2',6'-Dimethylphenoxy)phthalonitrile 99.0+%, TCI America™
SDP

Catalog No. D19835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19835G 5 g
D19831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19835G Supplier TCI America Supplier No. D19835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 221302-75-6
Molecular Formula C16H12N2O
Molecular Weight (g/mol) 248.285
MDL Number MFCD00059064
InChI Key LJUVUOSWARAIBZ-UHFFFAOYSA-N
PubChem CID 952073
IUPAC Name 4-(2,6-dimethylphenoxy)benzene-1,2-dicarbonitrile
SMILES CC1=C(C(=CC=C1)C)OC2=CC(=C(C=C2)C#N)C#N

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 5 g
Synonym 1,2-Dicyano-4-(2′,6′-dimethylphenoxy)benzene
Formula Weight 248.29
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2′,6′-Dimethylphenoxy)phthalonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.