Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(2,6-dioxo-1-propyl-1,2,3,6-tetrahydropurin-8-yl)benzene-1-sulfonic acid | 152529-79-8 | >=98.0% | 350.35 g/mol | C14H14N4O5S | 25 MG
SDP

Catalog No. 5000247492
Encompass_Preferred

PSB-1115 is a research-grade, selective A2B adenosine receptor antagonist used as a tool compound in pharmacology and receptor signaling studies.

  • Selective A2B adenosine receptor antagonist.
  • Intended for research and analytical applications.
  • CAS number 152529-79-8.
  • Molecular formula C14H14N4O5S.
  • Molecular weight 350.35 g/mol.
  • Recommended storage: -20°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied in small quantities suitable for analytical use.

Catalog No. 50-002-47492 Supplier Medchemexpress LLC Supplier No. HY10318225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSB-1115 is a research-grade, selective A2B adenosine receptor antagonist used as a tool compound in pharmacology and receptor signaling studies.

  • Selective A2B adenosine receptor antagonist.
  • Intended for research and analytical applications.
  • CAS number 152529-79-8.
  • Molecular formula C14H14N4O5S.
  • Molecular weight 350.35 g/mol.
  • Recommended storage: -20°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied in small quantities suitable for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.