Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(2,6-dioxo-1-propyl-1,2,3,6-tetrahydropurin-8-yl)benzene-1-sulfonic acid | 152529-79-8 | 99.0% | 350.35 | C14H14N4O5S | 50 MG
SDP

Catalog No. 5000246702
Encompass_Preferred

PSB-1115 is a selective A2B adenosine receptor antagonist supplied as a high-purity solid for research use. It is used in pharmacological and biochemical studies to investigate A2B receptor-mediated effects.

  • Selective A2B adenosine receptor antagonist
  • High purity (99.0%)
  • Solid form; available in multiple sizes including 50 MG
  • Molecular weight 350.35; formula C14H14N4O5S
  • CAS 152529-79-8

Catalog No. 50-002-46702 Supplier Medchemexpress LLC Supplier No. HY10318250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSB-1115 is a selective A2B adenosine receptor antagonist supplied as a high-purity solid for research use. It is used in pharmacological and biochemical studies to investigate A2B receptor-mediated effects.

  • Selective A2B adenosine receptor antagonist
  • High purity (99.0%)
  • Solid form; available in multiple sizes including 50 MG
  • Molecular weight 350.35; formula C14H14N4O5S
  • CAS 152529-79-8

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.