Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(2,6-dioxo-1-propyl-1,2,3,6-tetrahydropurin-8-yl)benzene-1-sulfonic acid | 152529-79-8 | 99.0% | 350.35 g/mol | C14H14N4O5S | 10 MG
SDP

Catalog No. 5000247491
Encompass_Preferred

PSB-1115 is a selective A2B adenosine receptor antagonist for pharmacological research. It inhibits TNBS-induced acetylcholine contraction in vitro and antagonizes NECA-induced inhibition of mucociliary clearance in vivo.

  • Selective A2B adenosine receptor antagonist
  • Purity 99.0%
  • Molecular weight 350.35 g/mol
  • Chemical formula C14H14N4O5S
  • CAS number 152529-79-8
  • Solubility: DMSO 100 mg/mL; H2O 10 mg/mL
  • Storage: -20°C; in solvent -80°C (6 months)
  • Intended for research use only

Catalog No. 50-002-47491 Supplier Medchemexpress LLC Supplier No. HY10318210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSB-1115 is a selective A2B adenosine receptor antagonist for pharmacological research. It inhibits TNBS-induced acetylcholine contraction in vitro and antagonizes NECA-induced inhibition of mucociliary clearance in vivo.

  • Selective A2B adenosine receptor antagonist
  • Purity 99.0%
  • Molecular weight 350.35 g/mol
  • Chemical formula C14H14N4O5S
  • CAS number 152529-79-8
  • Solubility: DMSO 100 mg/mL; H2O 10 mg/mL
  • Storage: -20°C; in solvent -80°C (6 months)
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.