Learn More
Medchemexpress LLC 4-(2-aminoethyl)-2-methoxyphenol | 554-52-9 | MFCD00060621 | >=97.0% | 167.21 | C9H13NO2 | 250 MG
SDP
Catalog No. 5000245044
Encompass_Preferred
3-Methoxytyramine is the O-methylated metabolite of dopamine (3-hydroxytyramine) that functions as a neuromodulator. Supplied as a solid analytical standard, it is intended for research and analytical applications including metabolic profiling and method development.
  • Analytical standard for research and analytical applications.
  • Molecular formula C9H13NO2; molecular weight 167.21 g/mol.
  • Available in small solid pack sizes suitable for laboratory assays.
  • Typical purity reported as ≥97% (verify batch certificate of analysis).
  • Used as a reference in metabolic studies and analytical workflows such as LC-MS.
Catalog No. 50-002-45044 Supplier Medchemexpress LLC Supplier No. HY103638A250MG
3-Methoxytyramine is the O-methylated metabolite of dopamine (3-hydroxytyramine) that functions as a neuromodulator. Supplied as a solid analytical standard, it is intended for research and analytical applications including metabolic profiling and method development.
  • Analytical standard for research and analytical applications.
  • Molecular formula C9H13NO2; molecular weight 167.21 g/mol.
  • Available in small solid pack sizes suitable for laboratory assays.
  • Typical purity reported as ≥97% (verify batch certificate of analysis).
  • Used as a reference in metabolic studies and analytical workflows such as LC-MS.