Learn More
Medchemexpress LLC 4-(2-aminoethyl)-7-(methylamino)chromen-2-one dihydrochloride | 1883548-88-6 | MFCD30182251 | 99.5% | 291.17 g/mol | C12H16Cl2N2O2 | 5 MG
SDP
Catalog No. 5000263663
Encompass_Preferred
FFN 206 dihydrochloride is a fluorescent vesicular monoamine transporter 2 (VMAT2) substrate used as a research probe to visualize and study monoamine uptake into synaptic vesicles. The compound is supplied as a stable dihydrochloride salt in small pack sizes for laboratory use.
  • Fluorescent VMAT2 substrate for visualization of vesicular uptake.
  • High purity suitable for analytical and preparative experiments.
  • Stable dihydrochloride salt with defined molecular weight and formula.
  • Recommended storage at -20°C; in solvent stability varies with temperature.
  • Available in small pack sizes for research use.
Catalog No. 50-002-63663 Supplier Medchemexpress LLC Supplier No. HY1103345MG
FFN 206 dihydrochloride is a fluorescent vesicular monoamine transporter 2 (VMAT2) substrate used as a research probe to visualize and study monoamine uptake into synaptic vesicles. The compound is supplied as a stable dihydrochloride salt in small pack sizes for laboratory use.
  • Fluorescent VMAT2 substrate for visualization of vesicular uptake.
  • High purity suitable for analytical and preparative experiments.
  • Stable dihydrochloride salt with defined molecular weight and formula.
  • Recommended storage at -20°C; in solvent stability varies with temperature.
  • Available in small pack sizes for research use.