Learn More
Medchemexpress LLC 4-(2′-aminoethyl)amino-1,8-dimethylimidazo[1,2-a]quinoxaline | 445430-58-0 | MFCD18089812 | 99.7% | 255.32 g·mol⁻1 | C14H17N5 | 25 MG
SDP
Catalog No. 5000243659
Encompass_Preferred
BMS-345541 is a small-molecule research reagent that selectively inhibits the catalytic subunits of IκB kinase (IKK). It binds an allosteric site on IKK and is used to probe NF-κB signaling pathways in biochemical and cellular studies. The compound is supplied with analytical characterization for research use only.
  • Selective inhibition of IKK catalytic subunits, with greater potency for IKK-2
  • Binds an allosteric site on the IKK enzyme
  • Reported IC50 values: IKK-2 = 0.3 μM; IKK-1 = 4 μM
  • High analytical purity suitable for research applications
  • Available in multiple pack sizes, including 25 MG
  • Molecular formula C14H17N5 and molecular weight 255.32 g·mol⁻1
  • Offered as solid and as ready-to-use DMSO solution formats
Catalog No. 50-002-43659 Supplier Medchemexpress LLC Supplier No. HY1051925MG
BMS-345541 is a small-molecule research reagent that selectively inhibits the catalytic subunits of IκB kinase (IKK). It binds an allosteric site on IKK and is used to probe NF-κB signaling pathways in biochemical and cellular studies. The compound is supplied with analytical characterization for research use only.
  • Selective inhibition of IKK catalytic subunits, with greater potency for IKK-2
  • Binds an allosteric site on the IKK enzyme
  • Reported IC50 values: IKK-2 = 0.3 μM; IKK-1 = 4 μM
  • High analytical purity suitable for research applications
  • Available in multiple pack sizes, including 25 MG
  • Molecular formula C14H17N5 and molecular weight 255.32 g·mol⁻1
  • Offered as solid and as ready-to-use DMSO solution formats