Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Aminoethyl)benzenesulfonamide, 99%, Thermo Scientific™

Catalog No. AAB2074522
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2074522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2074522 Supplier Thermo Scientific Chemicals Supplier No. B2074522

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35303-76-5
Molecular Formula C8H12N2O2S
Molecular Weight (g/mol) 200.256
MDL Number MFCD00010301
InChI Key FXNSVEQMUYPYJS-UHFFFAOYSA-N
Synonym 4-2-aminoethyl benzenesulfonamide, 4-2-aminoethyl benzene sulfonamide, 4-2-aminoethyl benzene-1-sulfonamide, 4-2-amino-ethyl-benzenesulfonamide, benzenesulfonamide, 4-2-aminoethyl, 4-2-aminoethyl benzenesulphonamide, chembl7087, 4-aminoethylbenzenesulfonamide, 2-4-sulfamoylphenyl ethylamine
PubChem CID 169682
IUPAC Name 4-(2-aminoethyl)benzenesulfonamide
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)N

Specifications

Melting Point 150°C to 152°C
Quantity 100 g
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 200.26
Percent Purity 99%
Chemical Name or Material 4-(2-Aminoethyl)benzenesulfonamide
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 252-501-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.