Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Aminoethyl)benzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. A136325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A136325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A136325G Supplier TCI America Supplier No. A136325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35303-76-5
Molecular Formula C8H12N2O2S
Molecular Weight (g/mol) 200.256
MDL Number MFCD00010301
InChI Key FXNSVEQMUYPYJS-UHFFFAOYSA-N
Synonym 4-2-aminoethyl benzenesulfonamide, 4-2-aminoethyl benzene sulfonamide, 4-2-aminoethyl benzene-1-sulfonamide, 4-2-amino-ethyl-benzenesulfonamide, benzenesulfonamide, 4-2-aminoethyl, 4-2-aminoethyl benzenesulphonamide, chembl7087, 4-aminoethylbenzenesulfonamide, 2-4-sulfamoylphenyl ethylamine
PubChem CID 169682
IUPAC Name 4-(2-aminoethyl)benzenesulfonamide
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)N

Specifications

Melting Point 149°C
Quantity 25 g
UN Number 3259
Formula Weight 200.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Aminoethyl)benzenesulfonamide
EINECSNumber (3)-4342
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.