Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(2-Aminoethyl)morpholine

Catalog No. A550045G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A550045G 5 g
A55004100G 100 g
A5500425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A550045G Supplier Sigma Aldrich Supplier No. A550045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-(2-aminoethyl)morpholine is widely used in biomedical applications as this moiety serves as an important lysosome-targeting group. some of its applications include:
  • Synthesis of the lysosome-targetable fluorescent probe for hydrogen sulfide imaging in living cells
  • Synthesis of 1,8-naphthalimide conjugated troger's bases as deoxyribonucleic acid (DNA) targeting fluorescent probe
  • Synthesis of intramolecular charge transfer-photoinduced electron transfer-fluorescence resonance energy transfer (ict-pet-fret) fluorescent probe for monitoring pH changes in living cells
  • It is also used as a precursor to synthesize a variety of antimicrobial agents

Specifications

Density 0.992 g/mL (at 25°C (literature))
Boiling Point 205°C (lit.)
Molecular Formula C6H14N2O
Refractive Index n20/D 1.476 (literature)
Linear Formula C6H14N2O
MDL Number MFCD00006182
Quantity 5 g
Synonym 2-Morpholinoethylamine
Molecular Weight (g/mol) 130.19
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.