Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Aminoethyl)morpholine, TCI America™
SDP

Catalog No. A0883100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0883100ML 100 mL
A0883500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0883100ML Supplier TCI America Supplier No. A0883100ML

Chemical Identifiers

CAS 2038-03-1
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.19
MDL Number MFCD00006182
InChI Key RWIVICVCHVMHMU-UHFFFAOYSA-N
Synonym 4-2-aminoethyl morpholine, 2-morpholinoethanamine, 4-morpholineethanamine, 2-morpholinoethylamine, n-aminoethylmorpholine, 2-morpholin-4-yl ethan-1-amine, morpholine, 4-2-aminoethyl, n-2-aminoethyl morpholine, n-2-aminoethylmorpholine, 2-morpholin-4-ylethylamine
PubChem CID 408285
IUPAC Name 2-morpholin-4-ylethanamine
SMILES C1COCCN1CCN

Specifications

Color White-Yellow
Boiling Point 205°C
Quantity 100 mL
UN Number 2923
Formula Weight 130.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-(2-Aminoethyl)morpholine
RTECSNumber QD7350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.