Learn More
Medchemexpress LLC 4-(2-azidoethyl)phenol | 74447-34-0 | MFCD24452536 | 99.8% | 163.18 g/mol | C8H9N3O | 10 MG
SDP
Catalog No. 5000295043
Encompass_Preferred
4-(2-Azidoethyl)phenol is an azide-containing phenol reagent used as a clickable probe for APEX2-mediated proximity labeling and downstream biotinylation. It supports reliable protein labeling for proteomic workflows and can be processed via multiple biotinylation strategies for analysis.
  • Clickable APEX2 probe for proximity labeling.
  • Enables two biotinylation routes for downstream analysis.
  • High purity suitable for biochemical and proteomic assays.
  • Available in multiple small pack sizes for experimental flexibility.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-95043 Supplier Medchemexpress LLC Supplier No. HY17317610MG
4-(2-Azidoethyl)phenol is an azide-containing phenol reagent used as a clickable probe for APEX2-mediated proximity labeling and downstream biotinylation. It supports reliable protein labeling for proteomic workflows and can be processed via multiple biotinylation strategies for analysis.
  • Clickable APEX2 probe for proximity labeling.
  • Enables two biotinylation routes for downstream analysis.
  • High purity suitable for biochemical and proteomic assays.
  • Available in multiple small pack sizes for experimental flexibility.
  • Stable when stored under recommended conditions.