Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Benzothiazolyl)aniline, 98%

Catalog No. AAH3305206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3305206 5 g
AAH3305203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3305206 Supplier Thermo Scientific Chemicals Supplier No. H3305206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6278-73-5 | C13H10N2S | 226.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6278-73-5
Molecular Formula C13H10N2S
Molecular Weight (g/mol) 226.30
MDL Number MFCD00182896
InChI Key WKRCOZSCENDENK-UHFFFAOYSA-N
Synonym 4-1,3-benzothiazol-2-yl aniline, 4-benzo d thiazol-2-yl aniline, 4-benzothiazol-2-yl-phenylamine, 2-4-aminophenyl benzothiazole, 4-1,3-benzothiazol-2-yl phenylamine, 4-benzothiazol-2-ylphenylamine, benzenamine, 4-2-benzothiazolyl, chembl93118, 4-1,3-benzothiazol-2-yl phenyl amine, benzenamine,4-2-benzothiazolyl
PubChem CID 234475
IUPAC Name 4-(1,3-benzothiazol-2-yl)aniline
SMILES NC1=CC=C(C=C1)C1=NC2=CC=CC=C2S1

Specifications

Melting Point 152°C to 154°C
Quantity 5 g
Formula Weight 226.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-(2-Benzothiazolyl)aniline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement

H301-H315-H319-H335

Toxic if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber CX9853100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.