Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(2-Bromoacetyl)benzonitrile | 20099-89-2 | 99.6% | C9H6BrNO | 25G
SDP

Catalog No. 5000357605
Encompass_Preferred

4-(2-Bromoacetyl)benzonitrile, also known as 4-Cyanophenacyl bromide, is a compound used in the synthesis of STA-5312, a potent and orally active microtubule inhibitor. This product is intended for research use only.

  • Can be utilized in the synthesis of STA-5312.
  • Appears as an off-white to yellow solid.
  • Store at 4°C, protected from light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in DMSO at 100 mg/mL.

Catalog No. 50-003-57605 Supplier Medchemexpress LLC Supplier No. 25GHY79638
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-(2-Bromoacetyl)benzonitrile, also known as 4-Cyanophenacyl bromide, is a compound used in the synthesis of STA-5312, a potent and orally active microtubule inhibitor. This product is intended for research use only.

  • Can be utilized in the synthesis of STA-5312.
  • Appears as an off-white to yellow solid.
  • Store at 4°C, protected from light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in DMSO at 100 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.