Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Chloro-4-nitrophenyl)morpholine, 97%, Thermo Scientific™

Catalog No. AAH63063MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63063MD 250 mg
AAH6306303 1 g
AAH6306306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63063MD Supplier Thermo Scientific Chemicals Supplier No. H63063MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55435-71-7
Molecular Formula C10H11ClN2O3
Molecular Weight (g/mol) 242.659
MDL Number MFCD00052898
InChI Key ZPKGWYXJULKUEH-UHFFFAOYSA-N
PubChem CID 108711
IUPAC Name 4-(2-chloro-4-nitrophenyl)morpholine
SMILES C1COCCN1C2=C(C=C(C=C2)[N+](=O)[O-])Cl

Specifications

Quantity 250 mg
Synonym 4-2-chloro-4-nitrophenyl morpholine, morpholine, 4-2-chloro-4-nitrophenyl, n-2-chloro-4-nitrophenyl morpholine, 4-2-chloro-4-nitro-phenyl-morpholine, 3-chloro-4-morpholino-1-nitrobenzene, maybridge1_003410, acmc-1b11p, morpholine,4-2-chloro-4-nitrophenyl, 3-chloro-4-morpholin-4-yl-1-nitro-benzene
Formula Weight 242.66
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(2-Chloro-4-nitrophenyl)morpholine
EINECSNumber 259-638-5
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.