Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-((2-Cyanopropan-2-yl)amino)-2-fluoro-N-methylbenzamide | 915087-32-0 | 99.1% | 235.26 | 250 MG
SDP

Catalog No. 5000402596
Encompass_Preferred

4-((2-Cyanopropan-2-yl)amino)-2-fluoro-N-methylbenzamide is a drug intermediate for the synthesis of various active compounds. It is for research use only.

  • Appearance: Solid
  • Purity: 99.1%
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-02596 Supplier Medchemexpress LLC Supplier No. 250MGHY20133
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-((2-Cyanopropan-2-yl)amino)-2-fluoro-N-methylbenzamide is a drug intermediate for the synthesis of various active compounds. It is for research use only.

  • Appearance: Solid
  • Purity: 99.1%
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.