Learn More
Medchemexpress LLC 4-[2-(cyclopropylmethoxy)-5-(methylsulfonyl)phenyl]-2-methyl-1(2H)-isoquinolinone | 1706738-98-8 | >98.0% | C21H21NO4S | 10MG
SDP
Catalog No. 5000259395
Encompass_Preferred
Trotabresib is an oral, potent, reversible small-molecule inhibitor of bromodomain and extra-terminal (BET) proteins. It is used in preclinical and clinical oncology research for advanced solid tumors and glioma and is supplied as a research-grade solid for translational studies.
  • Oral, reversible BET inhibitor suitable for epigenetic research.
  • Demonstrates antitumor activity in cell lines and xenograft models.
  • High purity (typically >98%) for reliable experimental results.
  • Solid powder form, soluble in common organic solvents such as DMSO.
  • Applicable to preclinical and clinical translational studies in oncology.
Catalog No. 50-002-59395 Supplier Medchemexpress LLC Supplier No. HY13757310MG
Trotabresib is an oral, potent, reversible small-molecule inhibitor of bromodomain and extra-terminal (BET) proteins. It is used in preclinical and clinical oncology research for advanced solid tumors and glioma and is supplied as a research-grade solid for translational studies.
  • Oral, reversible BET inhibitor suitable for epigenetic research.
  • Demonstrates antitumor activity in cell lines and xenograft models.
  • High purity (typically >98%) for reliable experimental results.
  • Solid powder form, soluble in common organic solvents such as DMSO.
  • Applicable to preclinical and clinical translational studies in oncology.