Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[2-(Dimethylamino)ethoxy]benzaldehyde, ≥95%, Thermo Scientific™

Catalog No. CC51504DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC51504DE 5 g
CC51504CB 250 mg
CC51504DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC51504DE Supplier Thermo Scientific Chemicals Supplier No. CC51504DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15182-92-0
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD07368978
InChI Key CBOKAZFQZOQTOC-UHFFFAOYSA-N
Synonym 4-2-dimethylamino ethoxy benzaldehyde, p-2-dimethylamino ethoxy benzaldehyde, 4-2-dimethylamino-ethoxy-benzaldehyde, pubchem12557, 4-2-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-ethoxy benzaldehyde, 4-2-dimethylaminoethyloxy benzaldehyde, 4-n,n-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-1-ethoxy benzaldehyde
PubChem CID 84823
IUPAC Name 4-[2-(dimethylamino)ethoxy]benzaldehyde
SMILES CN(C)CCOC1=CC=C(C=C1)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 193.24
Percent Purity 95%
Chemical Name or Material 4-[2-(Dimethylamino)ethoxy]benzaldehyde
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.