Learn More
Cayman Chemical 4-((2-FLUORO-3-((((6-METH 10MG
SDP
Supplier:  Cayman Chemical 2107410
Encompass_Preferred
CK-1827452 is a diaryl urea compound that increases cardiac myosin ATPase activity in a dose-dependent manner that is selective for cardiac myosin over non-cardiac myosins.1 It promotes myosin cross-bridge formation, increasing the duration and amount of myocyte contraction without affecting intracellular calcium or cAMP.1 It has been shown to increase cardiac efficiency in clinical studies of patients with chronic heart failure.
Catalog No. NC2007046