Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Hydroxy-3-isopropylaminopropoxy)phenylacetic acid, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014632 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4632 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4632 Supplier MilliporeSigma Supelco Supplier No. 7794725MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 56392-14-4
Linear Formula C14H21 NO4
MDL Number MFCD00437442
Synonym 4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetic acid; Atenolol acid; Metoprolol acid
Type Analytical Standard
Quantity 25 mg
Formula Weight 267.32
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.