Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Hydroxyethoxy)salicylic Acid 97.0+%, TCI America™
SDP

Catalog No. H07305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H07305G Supplier TCI America Supplier No. H07305G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 163451-82-9
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00143215
InChI Key INEIMLMPQGJCGK-UHFFFAOYSA-N
Synonym 2-Hydroxy-4-(2-hydroxyethoxy)benzoic Acid
PubChem CID 586463
IUPAC Name 2-hydroxy-4-(2-hydroxyethoxy)benzoic acid
SMILES C1=CC(=C(C=C1OCCO)O)C(=O)O

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 5 g
Formula Weight 198.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Hydroxyethoxy)salicylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.