Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Hydroxyethyl)benzonitrile, Thermo Scientific™

Catalog No. TL00139EA
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL00139EA 10 g
TL00139DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. TL00139EA Supplier Thermo Scientific Chemicals Supplier No. TL00139EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 69395-13-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
InChI Key RBSJBNYPTGMZIH-UHFFFAOYSA-N
Synonym 4-2-hydroxyethyl benzonitrile, benzonitrile, 4-2-hydroxyethyl, 2-4-cyanophenyl ethanol, 2-4-cyanophenyl ethan-1-one, 4-2-hydroxyethyl benzenecarbonitrile, 4-cyanobenzeneethanol, p-cyanophenethylalcohol, p-cyanophenethyl alcohol, 4-cyanophenethyl alcohol, acmc-20a4x6
PubChem CID 2800815
IUPAC Name 4-(2-hydroxyethyl)benzonitrile
SMILES C1=CC(=CC=C1CCO)C#N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CNC6H4(CH2)2OH
Quantity 10 g
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material 4-(2-Hydroxyethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.