Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Hydroxyhexafluoroisopropyl)benzoic acid, 97%

Catalog No. AAL1455403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1455403 1 g
AAL1455406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1455403 Supplier Thermo Scientific Chemicals Supplier No. L1455403
Add to Cart

CAS: 16261-80-6 | C10H6F6O3 | 288.15 g/mol

4-(2-Hydroxyhexafluoroisopropyl) benzoic acid is used in chemicals, pharmaceutical research, acts as a pharmaceutical intermediate and reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Hydroxyhexafluoroisopropyl) benzoic acid is used in chemicals, pharmaceutical research, acts as a pharmaceutical intermediate and reagent.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16261-80-6
Molecular Formula C10H6F6O3
Molecular Weight (g/mol) 288.15
MDL Number MFCD00236043
InChI Key DLJNNINHDYILFL-UHFFFAOYSA-N
Synonym 4-2-hydroxyhexafluoroisopropyl benzoic acid, 4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl benzoic acid, 4-perfluoro 2-hydroxyprop-2-yl benzoic acid, benzoic acid, 4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethyl, p-2-hydroxyhexafluoroisopropyl benzoic acid, 4-carboxyphenyl-1,1,1,3,3,3-hexafluoropropan-2-ol, 4-1,1,1,3,3,3-hexafluoro-2-hydroxyprop-2-yl benzoic acid, 4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethyl benzoic acid, 4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethyl benzoic acid #
PubChem CID 591169
IUPAC Name 4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)benzoic acid
SMILES OC(=O)C1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 122°C to 126°C
Odor Odorless
Quantity 1 g
Beilstein 2148278
Formula Weight 288.15
Percent Purity 97%
Chemical Name or Material 4-(2-Hydroxyhexafluoroisopropyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.