Learn More
Medchemexpress LLC 4-((2-(hydroxymethyl)phenyl)amino)-2-((6-methoxy-2-methyl-thiq-7-yl)amino)pyrimidine-5-carboxamide | 2739844-28-9 | 99.4% | C23H26N6O3 | 10MG
SDP
Catalog No. 5000261331
Encompass_Preferred
HPK1-IN-4 is a potent small-molecule inhibitor of hematopoietic progenitor kinase 1 (HPK1/MAP4K1) used as a preclinical immunotherapy research tool. It exhibits subnanomolar activity against HPK1 (reported IC50 0.061 nM), is supplied as an off-white to light yellow solid, and is characterized by high purity and a defined chemical identity suitable for in vitro and early-stage in vivo studies.
  • Potent HPK1 inhibition (IC50 0.061 nM).
  • High chemical purity suitable for research applications.
  • Well-characterized chemical identity and formula.
  • Solid small-molecule form for formulation flexibility.
  • Applicable as a preclinical tool to study HPK1-related signaling pathways.
Catalog No. 50-002-61331 Supplier Medchemexpress LLC Supplier No. HY13856910MG
HPK1-IN-4 is a potent small-molecule inhibitor of hematopoietic progenitor kinase 1 (HPK1/MAP4K1) used as a preclinical immunotherapy research tool. It exhibits subnanomolar activity against HPK1 (reported IC50 0.061 nM), is supplied as an off-white to light yellow solid, and is characterized by high purity and a defined chemical identity suitable for in vitro and early-stage in vivo studies.
  • Potent HPK1 inhibition (IC50 0.061 nM).
  • High chemical purity suitable for research applications.
  • Well-characterized chemical identity and formula.
  • Solid small-molecule form for formulation flexibility.
  • Applicable as a preclinical tool to study HPK1-related signaling pathways.