Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(2-Iodoethynyl)phenylboronic acid pinacol ester

Catalog No. RNI001111G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RNI001111G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RNI001111G Supplier Sigma Aldrich Supplier No. RNI001111G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-fluorophenyl isocyanate is the main component of essential oil from rhododendron thymifolium maxim
  • Molecular geometry of 2-fluorophenyl isocyanate has been investigated by microwave spectroscopy and nuclear magnetic resonance spectroscopy

Specifications

Molecular Formula C14H16BIO2
Linear Formula C14H16BIO2
MDL Number MFCD27978288
Quantity 5 g
Molecular Weight (g/mol) 353.99
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.