Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Keto-1-benzimidazolinyl)piperidine, 97%

Catalog No. AC156700250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156700250 25 g Glass bottle
AC156700010 1 g Glass bottle
AC156700050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC156700250 Supplier Thermo Scientific Chemicals Supplier No. 156700250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 20662-53-7 | C12H15N3O | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20662-53-7
Molecular Formula C12H15N3O
Molecular Weight (g/mol) 217.27
MDL Number MFCD00005714
InChI Key BYNBAMHAURJNTR-UHFFFAOYSA-N
Synonym 4-2-keto-1-benzimidazolinyl piperidine, 1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 4-2-keto-1-benzimidozolinyl piperidine, unii-4rp0aed2pc, 1-piperidin-4-yl-1h-benzimidazol-2-ol, 4rp0aed2pc, 1-4-piperidinyl-1,3-dihydro-2h-benzimidazol-2-one, 1-piperidin-4-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-piperidin-4-yl-3h-1,3-benzodiazol-2-one, 1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one
PubChem CID 88638
SMILES O=C1NC2=CC=CC=C2N1C1CCNCC1

Specifications

Melting Point 183.0°C to 185.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: 32g/100mL methanol- 13g/100mL ethanol,7g/100mL 2-propanol- 0.75g/100mL acetone,3.1g/100mL methylene chloride- 9.4g/100mL n,n-dmf
IUPAC Name 3-piperidin-4-yl-1H-benzimidazol-2-one
Formula Weight 217.27
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-(2-Keto-1-benzimidazolinyl)piperidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 243-950-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.