Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(2-Methoxyphenyl)pyrimidin-2-amine

Catalog No. JRD09941G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD09941G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD09941G Supplier Sigma Aldrich Supplier No. JRD09941G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Benzocyclobutene was used as the adhesive bonding material in adhesive wafer-level bonding technique
  • Photosensitive benzocyclobutene has been widely used as dielectric material in multilayer packaging applications
  • It was used in fabrication of high density multilayer interconnect structures using gold metallization and polymer interlayer dielectric

Specifications

CAS 915070-01-8
Molecular Formula C11H11N3O
Linear Formula C11H11N3O
MDL Number MFCD07381837
Quantity 500 mg
Molecular Weight (g/mol) 201.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.