Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(2-Methyl-4-thiazolyl)phenol | 30686-73-8 | 98.7% | 250 MG
SDP

Catalog No. 5000414365
Encompass_Preferred

4-(2-Methyl-4-thiazolyl)phenol is a biochemical reagent suitable for life science related research as a biological material or organic compound. It is intended for research use only.

  • Ships at room temperature (continental US).
  • Store as solid at 4°C under nitrogen.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month under nitrogen.
  • Molecular weight of 191.25.
  • Chemical formula C10H9NOS.
  • Appears as a white to off-white solid.
  • Soluble in DMSO (50 mg/mL).

Catalog No. 50-004-14365 Supplier Medchemexpress LLC Supplier No. 250MGHYD0126
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-(2-Methyl-4-thiazolyl)phenol is a biochemical reagent suitable for life science related research as a biological material or organic compound. It is intended for research use only.

  • Ships at room temperature (continental US).
  • Store as solid at 4°C under nitrogen.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month under nitrogen.
  • Molecular weight of 191.25.
  • Chemical formula C10H9NOS.
  • Appears as a white to off-white solid.
  • Soluble in DMSO (50 mg/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.