Learn More
Medchemexpress LLC 4-[2-(methylthio)phenyl]-N-(1,2,3,4-tetrahydro-1-naphthalenyl)-1-piperazinehexanamide, monohydrochloride | 824958-12-5 | 98.1% | 488.13 | C27H38ClN3OS | 1 ML
SDP
Catalog No. 5000247212
Encompass_Preferred
LP44 hydrochloride is the hydrochloride salt of LP44, a potent and selective 5-HT7 (serotonin 7) receptor agonist used as a research tool in neuroscience to probe receptor pharmacology and physiological responses.
  • Selective 5-HT7 receptor agonist with nanomolar affinity.
  • Available as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid forms.
  • High purity (~98.1%), suitable for research applications.
  • Molecular formula C27H38ClN3OS; molecular weight 488.13.
  • Supplied as a hydrochloride salt to improve solubility in DMSO.
Catalog No. 50-002-47212 Supplier Medchemexpress LLC Supplier No. HY10310110MM 1ML
LP44 hydrochloride is the hydrochloride salt of LP44, a potent and selective 5-HT7 (serotonin 7) receptor agonist used as a research tool in neuroscience to probe receptor pharmacology and physiological responses.
  • Selective 5-HT7 receptor agonist with nanomolar affinity.
  • Available as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid forms.
  • High purity (~98.1%), suitable for research applications.
  • Molecular formula C27H38ClN3OS; molecular weight 488.13.
  • Supplied as a hydrochloride salt to improve solubility in DMSO.