Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[4-(2-Morpholin-4-ylethoxy)phenyl]methanol, 97%, Thermo Scientific™

Catalog No. CC42109CB
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC42109CB 250 mg
CC42109DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC42109CB Supplier Thermo Scientific Chemicals Supplier No. CC42109CB

Chemical Identifiers

CAS 852180-76-8
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.30
MDL Number MFCD07368553
InChI Key LZRLXLXSGAEQLA-UHFFFAOYSA-N
Synonym 4-2-morpholin-4-ylethoxy phenyl methanol, 4-2-morpholin-4-yl ethoxy phenyl methanol, 4-2-morpholinoethoxy phenyl methanol, benzenemethanol,4-2-4-morpholinyl ethoxy, 4-2-morpholin-4-yl-ethoxy-phenyl-methanol, 4-2-morpholin-4-ylethoxy phenyl methan-1-ol
PubChem CID 7060580
IUPAC Name [4-(2-morpholin-4-ylethoxy)phenyl]methanol
SMILES OCC1=CC=C(OCCN2CCOCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 237.3
Percent Purity 97%
Chemical Name or Material [4-(2-Morpholin-4-ylethoxy)phenyl]methanol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.