Learn More
Medchemexpress LLC 4-(2-((N-(4-chlorobenzyl)-2,3,4,5,6-pentafluorophenyl)sulfonamido)-N-(3,5-di-tert-butylbenzyl)acetamido) benzoic acid | 1834571-82-2 | MFCD34469333 | 99.9% | 751.20 | C37H36ClF5N2O5S | 10MM 1ML
SDP
Catalog No. 5000236908
Encompass_Preferred
AC-4-130 is a research-grade small-molecule inhibitor that targets the STAT5 SH2 domain, disrupting STAT5 activation, dimerization, nuclear translocation, and downstream transcription. It is supplied for in vitro biochemical and cellular research in multiple solid masses and as a 10 mM solution in DMSO.
  • Potent inhibitor of the STAT5 SH2 domain.
  • Disrupts STAT5 activation, dimerization, and nuclear translocation.
  • High purity (>99.9%) suitable for analytical and biological studies.
  • Available as a 10 mM DMSO solution or in multiple solid pack sizes for dosing flexibility.
  • Characterized molecular weight and formula for compound confirmation.
Catalog No. 50-002-36908 Supplier Medchemexpress LLC Supplier No. HY12450010MM 1ML
AC-4-130 is a research-grade small-molecule inhibitor that targets the STAT5 SH2 domain, disrupting STAT5 activation, dimerization, nuclear translocation, and downstream transcription. It is supplied for in vitro biochemical and cellular research in multiple solid masses and as a 10 mM solution in DMSO.
  • Potent inhibitor of the STAT5 SH2 domain.
  • Disrupts STAT5 activation, dimerization, and nuclear translocation.
  • High purity (>99.9%) suitable for analytical and biological studies.
  • Available as a 10 mM DMSO solution or in multiple solid pack sizes for dosing flexibility.
  • Characterized molecular weight and formula for compound confirmation.