Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Naphthyl)phenol, 98%, Thermo Scientific™

Catalog No. AAH6433203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6433203 1 g
AAH6433206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6433203 Supplier Thermo Scientific Chemicals Supplier No. H6433203

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6336-82-9
Molecular Formula C16H12O
Molecular Weight (g/mol) 220.27
MDL Number MFCD18312886
InChI Key NIRHUNSXEDESLN-UHFFFAOYSA-N
Synonym 4-naphthalen-2-yl phenol, 4-naphthalen-2-yl-phenol, 4-2-naphthyl phenol, d0l3ax, 2-4-hydroxyphenyl naphthalene
PubChem CID 236868
SMILES OC1=CC=C(C=C1)C1=CC=C2C=CC=CC2=C1

Specifications

Quantity 1 g
Beilstein 2524079
IUPAC Name 4-(naphthalen-2-yl)phenol
Formula Weight 220.27
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(2-Naphthyl)phenol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.