Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Oxo-1-pyrrolidinyl)benzaldehyde, 97%, Thermo Scientific™

Catalog No. p-3075534
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO01193DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01193DA Supplier Thermo Scientific Chemicals Supplier No. MO01193DA
Add to Cart

Chemical Identifiers

CAS 36151-45-8
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.214
MDL Number MFCD00454011
InChI Key VADZUJOCSAESJS-UHFFFAOYSA-N
Synonym 4-2-oxopyrrolidin-1-yl benzaldehyde, 4-2-oxo-1-pyrrolidinyl benzaldehyde, 4-2-oxo-pyrrolidin-1-yl-benzaldehyde, 4-2-oxopyrrolidinyl benzaldehyde, benzaldehyde,4-2-oxo-1-pyrrolidinyl, 4-2-oxo-pyrrolidin-1-yl benzaldehyde, 4-2-oxidanylidenepyrrolidin-1-yl benzaldehyde
PubChem CID 2794706
IUPAC Name 4-(2-oxopyrrolidin-1-yl)benzaldehyde
SMILES C1CC(=O)N(C1)C2=CC=C(C=C2)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material 4-(2-Oxo-1-pyrrolidinyl)benzaldehyde
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.