Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Pyridylazo)resorcinol, ≥95.0%, Solstice

Catalog No. 60047646 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-646 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-646 Supplier Solstice Supplier No. 829705G

missing translation for 'validMainframeMsg'

for the spectrophotometric det. of Hg, indicator for complexometric titration,≥95.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1141-59-9
Molecular Formula C11H9N3O2
Molecular Weight (g/mol) 215.212
MDL Number MFCD00006256
InChI Key VLCAILLZPUINNF-LCYFTJDESA-N
PubChem CID 5474737
IUPAC Name (4Z)-3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one
SMILES C1=CC=NC(=C1)NN=C2C=CC(=O)C=C2O

Specifications

Melting Point 192°C to 202°C
UN Number NONH for all modes of transport
Beilstein 184665
Formula Weight 215.21g/mol
Percent Purity ≥95.0%
Packaging Glass bottle
Quantity 5 g
Chemical Name or Material 4-(2-Pyridylazo)resorcinol
Hazard Statement P261-P305 + P351 + P338
RTECSNumber 214-528-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.