Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Pyridylazo)Resorcinol Monosodium Salt Hydrate, Sigma-Aldrich

Catalog No. 01258993 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-993 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-993 Supplier MilliporeSigma Supelco Supplier No. 17826810G
Add to Cart
Add to Cart

4-(2-Pyridylazo)resorcinol monosodium salt hydrate has been used in the preparation of titanium reagent for the spectrometric measurement of hydrogen peroxide (H2O2) and superoxide anion (O2-). It is also suitable to measure the concentration of Zn2+, Zinc transition metal ions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16593-81-0
Molecular Weight (g/mol) 237.19 g/mol

Specifications

Physical Form Powder, Crystals and/or Chunks
Quantity 10 g
Percent Purity ≥97.5%
Packaging Amber bottle
Chemical Name or Material PAR monosodium salt hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.