Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Pyridylazo)resorcinol monosodium salt monohydrate

Catalog No. AAA1810303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1810303 1 g
AAA1810306 5 g
AAA1810314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1810303 Supplier Thermo Scientific Chemicals Supplier No. A1810303
Add to Cart

CAS: 16593-81-0 | C11H9N3O2 | 215.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16593-81-0
Molecular Formula C11H9N3O2
Molecular Weight (g/mol) 215.21
MDL Number MFCD00150712,MFCD00150712,MFCD00150712,MFCD00006256
InChI Key VLCAILLZPUINNF-UKTHLTGXSA-N
Synonym 4-2-pyridylazo resorcinol monosodium saltmonohydrate
PubChem CID 132988475
IUPAC Name 3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one;sodium;hydrate
SMILES OC1=CC(=O)C=C\C1=N/NC1=CC=CC=N1

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 3825637
Formula Weight 255.21 (237.19 Anhydrous)
Chemical Name or Material 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-339-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.