Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Pyridylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog No. AAH52843MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52843MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH52843MD Supplier Thermo Scientific Chemicals Supplier No. H52843MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(2-Pyridylcarbamoyl)benzeneboronic acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850568-25-1
Molecular Formula C12H11BN2O3
Molecular Weight (g/mol) 242.041
MDL Number MFCD06659876
InChI Key LEAZDTWQWNXKJM-UHFFFAOYSA-N
Synonym 4-pyridin-2-yl aminocarbonylphenylboronic acid, 4-pyridin-2-yl-aminocarbonyl benzeneboronic acid, 4-pyridin-2-ylcarbamoyl phenyl boronic acid, 4-2-pyridylcarbamoyl benzeneboronic acid, 4-pyridin-2-yl carbamoyl phenylboronic acid, acmc-209q1n, n-2-pyridinyl 4-boronobenzamide, 4-pyridin-2-ylcarbamoyl phenylboronic acid, 4-pyridin-2-yl carbamoyl phenyl boronic acid, 4-2-pyridylaminocarbonyl phenylboronic acid
PubChem CID 44119539
IUPAC Name [4-(pyridin-2-ylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NC2=CC=CC=N2)(O)O

Specifications

Melting Point 124°C to 128°C
Quantity 250 mg
Formula Weight 242.04
Percent Purity 97%
Chemical Name or Material 4-(2-Pyridylcarbamoyl)benzeneboronic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.