Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(2-pyridyldithio)butanoic acid | 250266-79-6 | >98.0% | 229.32 g·mol⁻¹ | C9H11NO2S2 | 250 MG
SDP

Catalog No. 5000202682
Encompass_Preferred

Acid-C3-SSPy is a cleavable disulfide linker used as a building block in the synthesis of DBA-DM4, an agent-linker conjugate for antibody-drug conjugates (ADC). It is supplied as a solid research reagent and is intended for laboratory use only.

  • Cleavable disulfide linker suitable for ADC synthesis
  • Used as a building block in preparing agent-linker conjugates
  • Molecular formula: C9H11NO2S2
  • Molecular weight: 229.32 g·mol⁻¹
  • CAS number: 250266-79-6
  • Purity: >98.0%
  • Supplied as a 250 mg solid

Catalog No. 50-002-02682 Supplier Medchemexpress LLC Supplier No. HY141597250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acid-C3-SSPy is a cleavable disulfide linker used as a building block in the synthesis of DBA-DM4, an agent-linker conjugate for antibody-drug conjugates (ADC). It is supplied as a solid research reagent and is intended for laboratory use only.

  • Cleavable disulfide linker suitable for ADC synthesis
  • Used as a building block in preparing agent-linker conjugates
  • Molecular formula: C9H11NO2S2
  • Molecular weight: 229.32 g·mol⁻¹
  • CAS number: 250266-79-6
  • Purity: >98.0%
  • Supplied as a 250 mg solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.